Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272011.1 | NP_001258940.1 | 11 | 1387 | ||
NM_014226.2 | NP_055041.1 | 12 | 1477 | |||
NR_073541.1 | 6 | 757 | ||||
NR_073542.1 | 5 | 710 | ||||
NR_073543.1 | 5 | 673 | ||||
XM_006720226.2 | XP_006720289.1 | 10 | 1452 | |||
XM_011537053.2 | XP_011535355.1 | 5 | 1594 | |||
XM_017021549.1 | XP_016877038.1 | 12 | 1474 | |||
XM_017021550.1 | XP_016877039.1 | 11 | 1373 | |||
XM_017021551.1 | XP_016877040.1 | 13 | 1417 | |||
XM_017021552.1 | XP_016877041.1 | 11 | 1605 | |||
XM_017021553.1 | XP_016877042.1 | 10 | 1312 | |||
XM_017021554.1 | XP_016877043.1 | 10 | 1311 | |||
XM_017021557.1 | XP_016877046.1 | 9 | 1352 | |||
XM_017021558.1 | XP_016877047.1 | 9 | 1315 | |||
XM_017021559.1 | XP_016877048.1 | 11 | 1466 | |||
GenBank mRNA | AB022694.1 | - | 12 | 1471 | ||
AK094003.1 | - | 2 | 1849 | |||
AK094747.1 | - | 9 | 2259 | |||
AK131542.1 | - | 2 | 3332 | |||
AK302349.1 | - | 11 | 1372 | |||
AK312778.1 | - | 12 | 1468 | |||
BC026069.1 | - | 11 | 1888 | 4838296 | ||
BC043179.1 | - | 10 | 2233 | 5287879 | ||
BC053536.1 | - | 11 | 2432 | 5175851 | ||
U46191.1 | - | 5 | 650 | |||
U46192.1 | - | 5 | 700 | |||
U46193.1 | - | 6 | 757 |