Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000324.2 | NP_000315.2 | 5 | 753 | ||
GenBank mRNA | AB938314.1 | - | 5 | 691 | ||
AF031548.1 | - | 5 | 691 | |||
AF031549.1 | - | 5 | 718 | |||
AF178841.1 | - | 5 | 718 | |||
AF179682.1 | - | 5 | 718 | |||
AF179684.1 | - | 5 | 718 | |||
AF179685.1 | - | 5 | 718 | |||
AF187847.1 | - | 5 | 718 | |||
AK313505.1 | - | 5 | 753 | |||
BC012605.1 | - | 5 | 722 | 4063377 | ||
KJ543706.1 | - | 5 | 691 | |||
X64594.1 | - | 5 | 718 |