Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256820.1 | NP_001243749.1 | 4 | 363 | ||
NM_001256821.1 | NP_001243750.1 | 5 | 390 | |||
NM_006912.5 | NP_008843.1 | 5 | 512 | |||
GenBank mRNA | AF084462.1 | - | 5 | 423 | ||
AK298768.1 | - | 4 | 363 | |||
AK314239.1 | - | 5 | 488 | |||
BC104186.2 | - | 5 | 383 | 40030939 | ||
BC104187.2 | - | 5 | 383 | 40030940 | ||
BI916790.1 | - | 5 | 373 | |||
BM546985.1 | - | 5 | 506 | |||
BQ918446.1 | - | 5 | 390 | |||
CR407639.1 | - | 4 | 309 | |||
U71203.1 | - | 5 | 444 | |||
U78165.1 | - | 5 | 444 | |||
Y07566.1 | - | 5 | 454 |