Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005406.2 | NP_005397.1 | 3, 4 | 1207 | ||
GenBank mRNA | AK057038.1 | - | 3, 4 | 1208 | ||
AK307388.1 | - | 3, 4 | 1022 | |||
BC113114.1 | - | 3, 4 | 282 | 7961989 | ||
U43195.1 | - | 3, 4 | 266 |