Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006269.1 | NP_006260.1 | 4 | 4640 | ||
XM_017013721.1 | XP_016869210.1 | 9 | 5803 | |||
XM_017013722.1 | XP_016869211.1 | 4 | 4656 | |||
GenBank mRNA | AF141021.1 | - | 4 | 4640 | ||
AF146592.1 | - | 4 | 4588 |