Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037535.2 | NP_001032624.1 | 5 | 873 | ||
NM_001037536.2 | NP_001032625.1 | 5 | 870 | |||
NM_001037540.2 | NP_001032629.1 | 7 | 1103 | |||
NM_006746.5 | NP_006737.2 | 6 | 1022 | |||
XM_005274578.2 | XP_005274635.1 | 7 | 1098 | |||
XM_005274579.2 | XP_005274636.1 | 6 | 1011 | |||
XM_006724508.3 | XP_006724571.1 | 6 | 4003 | |||
XM_006724509.3 | XP_006724572.1 | 5 | 3919 | |||
XM_011545564.2 | XP_011543866.1 | 6 | 4006 | |||
XM_017029723.1 | XP_016885212.1 | 5 | 3922 | |||
GenBank mRNA | AF160728.1 | - | 5 | 810 | ||
AK313993.1 | - | 5 | 831 | |||
BC026159.2 | - | 6 | 1002 | 4827364 | ||
BC105028.1 | - | 3 | 479 | 8144031 | ||
BX647757.1 | - | 4 | 2787 |