Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002999.3 | NP_002990.2 | 3, 4 | 286 | ||
XM_011528977.2 | XP_011527279.1 | 2, 3 | 141 | |||
GenBank mRNA | AK222695.1 | - | 3, 4 | 286 | ||
AK223243.1 | - | 3, 4 | 288 | |||
AK303964.1 | - | 2, 3 | 147 | |||
AK312507.1 | - | 3, 4 | 286 | |||
BC030805.1 | - | 3, 4 | 286 | 4690309 | ||
CR542045.1 | - | 3, 4 | 246 | |||
CR542074.1 | - | 3, 4 | 246 | |||
D13292.1 | - | 3, 4 | 286 | |||
X67016.1 | - | 3, 4 | 272 |