Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006923.3 | NP_008854.2 | 1 | 308 | ||
XM_011525106.2 | XP_011523408.1 | 2 | 165 | |||
GenBank mRNA | AK310890.1 | - | 1 | 40 | ||
BC000500.2 | - | 1 | 23 | 2821329 | ||
BC001406.2 | - | 1 | 30 | 3140716 | ||
BQ056236.1 | - | 1 | 308 | |||
BT006709.1 | - | 1 | 12 | |||
CR456708.1 | - | 1 | 12 | |||
D50645.1 | - | 1 | 51 |