Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005066.2 | NP_005057.1 | 3 | 1366 | ||
NR_136702.1 | 3 | 1369 | ||||
NR_136703.1 | 3 | 1369 | ||||
XM_005271112.4 | XP_005271169.1 | 3 | 1385 | |||
XM_005271113.4 | XP_005271170.1 | 3 | 1385 | |||
XM_005271115.4 | XP_005271172.1 | 3 | 1385 | |||
XM_011541950.2 | XP_011540252.1 | 3 | 1385 | |||
XM_017002053.1 | XP_016857542.1 | 3 | 1385 | |||
XM_017002054.1 | XP_016857543.1 | 3 | 1385 | |||
GenBank mRNA | AK307985.1 | - | 4 | 1174 | ||
BC004534.2 | - | 3 | 1054 | |||
BC027708.2 | - | 3 | 1357 | |||
BC051192.1 | - | 3 | 1368 | 5262885 | ||
X16850.1 | - | 2 | 340 | |||
X70944.1 | - | 3 | 1358 |