Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005626.4 | NP_005617.2 | 6 | 1219 | ||
XM_011541951.2 | XP_011540253.1 | 5 | 929 | |||
GenBank mRNA | AB209856.1 | - | 8 | 7049 | ||
AK223444.1 | - | 6 | 996 | |||
AK291509.1 | - | 6 | 941 | |||
AK293706.1 | - | 4 | 687 | |||
BC002781.2 | - | 6 | 943 | 3619538 | ||
BT007415.1 | - | 6 | 846 | |||
L14076.1 | - | 6 | 893 |