Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000023.3 | NP_000014.1 | 2 | 186 | ||
NM_001135697.2 | NP_001129169.1 | 2 | 186 | |||
NR_135553.1 | 2 | 186 | ||||
XM_011525120.2 | XP_011523422.2 | 2 | 652 | |||
XM_011525121.2 | XP_011523423.2 | 2 | 654 | |||
XM_011525122.2 | XP_011523424.2 | 2 | 652 | |||
XM_011525123.2 | XP_011523425.2 | 2 | 652 | |||
XM_011525124.2 | XP_011523426.1 | 2 | 162 | |||
XM_017024950.1 | XP_016880439.1 | 2 | 648 | |||
GenBank mRNA | AK290622.1 | - | 2 | 159 | ||
AK293614.1 | - | 2 | 162 | |||
AK300710.1 | - | 2 | 168 | |||
BC025702.1 | - | 2 | 158 | 5222483 | ||
DC306167.1 | - | 2 | 245 | |||
KU178331.1 | - | 2 | 130 | |||
KU178332.1 | - | 2 | 130 | |||
L34355.1 | - | 2 | 167 | |||
L35853.1 | - | 2 | 167 | |||
L46810.1 | - | 2 | 167 | |||
U08895.1 | - | 2 | 141 |