Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003022.2 | NP_003013.1 | 2 | 476 | ||
XM_011531013.1 | XP_011529315.1 | 3 | 280 | |||
XM_011531014.1 | XP_011529316.1 | 4 | 599 | |||
GenBank mRNA | AF042081.1 | - | 2 | 157 | ||
AK024892.1 | - | 3 | 250 | |||
AK129610.1 | - | 2 | 161 | |||
AK309350.1 | - | 4 | 605 | |||
AK309351.1 | - | 3 | 351 | |||
AK309352.1 | - | 3 | 371 | |||
AK310011.1 | - | 3 | 314 | |||
AK310155.1 | - | 3 | 269 | |||
AK315897.1 | - | 2 | 141 | |||
AL136718.1 | - | 2 | 177 | |||
BC016709.1 | - | 2 | 182 | 4105289 | ||
BC103762.1 | - | 2 | 301 | 6179172 | ||
CR533478.1 | - | 2 | 79 | |||
DA487377.1 | - | 2 | 476 | |||
FJ224348.1 | - | 2 | 157 |