Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166356.1 | NP_001159828.1 | 9, 10 | 1291 | ||
NM_001166357.1 | NP_001159829.1 | 9, 10 | 1383 | |||
NM_001166358.1 | NP_001159830.1 | 9, 10 | 1282 | |||
NM_001166359.1 | NP_001159831.1 | 9, 10 | 1175 | |||
NM_005412.5 | NP_005403.2 | 9, 10 | 1321 | |||
NR_029415.1 | 8, 9 | 1123 | ||||
NR_029416.1 | 8, 9 | 1202 | ||||
NR_029417.1 | 8, 9 | 1241 | ||||
NR_048562.1 | 8, 9 | 1331 | ||||
XM_011538675.2 | XP_011536977.1 | 9, 10 | 1261 | |||
XM_011538676.2 | XP_011536978.1 | 9, 10 | 1252 | |||
XM_011538677.2 | XP_011536979.1 | 8, 9 | 1162 | |||
XM_011538678.2 | XP_011536980.1 | 8, 9 | 1144 | |||
GenBank mRNA | AK055053.1 | - | 4, 5 | 2524 | ||
AK223555.1 | - | 9, 10 | 1183 | |||
AK295941.1 | - | 8, 9 | 1177 | |||
AK296183.1 | - | 9, 10 | 1135 | |||
AK297173.1 | - | 8, 9 | 1083 | |||
AK298231.1 | - | 8, 9 | 976 | |||
AK301339.1 | - | 9, 10 | 961 | |||
AK301443.1 | - | 8, 9 | 1145 | |||
AK303827.1 | - | 8, 9 | 997 | |||
AK315916.1 | - | 9, 10 | 1375 | |||
BC011911.2 | - | 9, 10 | 1125 | 4139001 | ||
BC013677.2 | - | 9, 10 | 1185 | 3863368 | ||
BC025355.1 | - | 6, 7 | 708 | |||
BC032584.1 | - | 9, 10 | 1164 | 5499481 | ||
BC044211.1 | - | 9, 10 | 1161 | 5521082 | ||
BC091501.1 | - | 8, 9 | 1045 | |||
BT006866.1 | - | 9, 10 | 1115 | |||
BX647711.1 | - | 9, 10 | 1242 | |||
BX648847.1 | - | 7, 8 | 723 | |||
EU668344.1 | - | 9, 10 | 1125 | |||
KU178333.1 | - | 9, 10 | 1115 | |||
KU178334.1 | - | 8, 9 | 751 | |||
L11932.1 | - | 8, 9 | 1026 |