Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SHMT2 SIRNA Assays ›
Gene Symbol:
SHMT2
Gene Name:
serine hydroxymethyltransferase 2
Gene Aliases:
GLYA, HEL-S-51e, SHMT
Chromosome Location:
Chr.12: 57229573 - 57234935 on Build GRCh38
UniGene ID:
Hs.741179
Species:
Human
siRNA ID
HSS109734
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SHMT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166356.1
NP_001159828.1
11
1538
NM_001166357.1
NP_001159829.1
11
1630
NM_001166358.1
NP_001159830.1
11
1529
NM_001166359.1
NP_001159831.1
11
1422
NM_005412.5
NP_005403.2
11
1568
NR_029415.1
10
1370
NR_029416.1
10
1449
NR_029417.1
10
1488
NR_048562.1
10
1578
XM_011538675.2
XP_011536977.1
11
1508
XM_011538676.2
XP_011536978.1
11
1499
XM_011538677.2
XP_011536979.1
10
1409
XM_011538678.2
XP_011536980.1
10
1391
GenBank mRNA
AK055053.1
-
6
2771
AK223555.1
-
11
1430
AK295941.1
-
10
1424
AK296183.1
-
11
1382
AK297173.1
-
10
1330
AK298231.1
-
10
1223
AK301339.1
-
11
1208
AK301443.1
-
10
1392
AK303827.1
-
10
1244
AK315916.1
-
11
1622
BC011911.2
-
11
1372
4139001
BC013677.2
-
11
1432
3863368
BC025355.1
-
8
955
BC032584.1
-
11
1411
5499481
BC044211.1
-
11
1408
5521082
BC091501.1
-
10
1292
BT006866.1
-
11
1362
BX647711.1
-
11
1489
BX648847.1
-
9
970
EU668344.1
-
11
1372
KU178333.1
-
11
1362
KU178334.1
-
10
998
L11932.1
-
10
1273
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
glycine metabolic process
L-serine metabolic process
L-serine biosynthetic process
one-carbon metabolic process
positive regulation of cell proliferation
glycine biosynthetic process from serine
response to type I interferon
tetrahydrofolate interconversion
tetrahydrofolate metabolic process
folic acid metabolic process
protein tetramerization
protein homotetramerization
protein K63-linked deubiquitination
chromatin binding
glycine hydroxymethyltransferase activity
protein binding
L-allo-threonine aldolase activity
amino acid binding
pyridoxal phosphate binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0