Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190997.2 | NP_001177926.1 | 12, 13 | 1500 | ||
NM_016615.4 | NP_057699.2 | 14, 15 | 1776 | |||
XM_006719008.3 | XP_006719071.1 | 9, 10 | 1563 | |||
XM_011521012.2 | XP_011519314.1 | 14, 15 | 1844 | |||
XM_011521013.1 | XP_011519315.1 | 15, 16 | 1912 | |||
XM_011521014.1 | XP_011519316.1 | 13, 14 | 1668 | |||
XM_017019842.1 | XP_016875331.1 | 10, 11 | 1661 | |||
XM_017019843.1 | XP_016875332.1 | 10, 11 | 1259 | |||
XM_017019845.1 | XP_016875334.1 | 10, 11 | 1161 | |||
GenBank mRNA | AK296127.1 | - | 12, 13 | 1463 | ||
AK296209.1 | - | 15, 16 | 1707 | |||
AK313511.1 | - | 14, 15 | 1732 | |||
BC022392.1 | - | 14, 15 | 1739 | 4612245 | ||
DB540466.1 | - | 1, 2 | 40 | |||
U76343.2 | - | 14, 15 | 1620 |