Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000338.2 | NP_000329.2 | 15 | 2136 | ||
NM_001184832.1 | NP_001171761.1 | 15 | 2136 | |||
XM_005254606.1 | XP_005254663.1 | 15 | 2136 | |||
GenBank mRNA | AK295375.1 | - | 14 | 1589 | ||
AK298275.1 | - | 10 | 1355 | |||
AK298312.1 | - | 15 | 2135 | |||
BX647067.1 | - | 10 | 6059 | |||
BX647484.1 | - | 11 | 6043 | |||
EF559316.2 | - | 14 | 1934 | |||
U58130.1 | - | 14 | 1939 |