Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145961.1 | NP_001139433.1 | 6 | 700 | ||
NM_001145962.1 | NP_001139434.1 | 5 | 605 | |||
NM_001145963.1 | NP_001139435.1 | 6 | 579 | |||
NM_001145964.1 | NP_001139436.1 | 6 | 570 | |||
NM_005072.4 | NP_005063.1 | 6 | 700 | |||
GenBank mRNA | AF047338.1 | - | 6 | 559 | ||
AF054505.1 | - | 6 | 559 | |||
AF054506.1 | - | 6 | 559 | |||
AK097808.1 | - | 7 | 661 | |||
AK293906.1 | - | 6 | 609 | |||
AK293956.1 | - | 5 | 495 | |||
AK299042.1 | - | 6 | 486 | |||
AK302790.1 | - | 6 | 579 | |||
AK311073.1 | - | 6 | 600 | |||
AK316415.1 | - | 6 | 560 | |||
BC021193.2 | - | 6 | 628 | 3349710 | ||
BC035480.1 | - | 6 | 1881 | |||
U55054.1 | - | 6 | 614 |