Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003074.3 | NP_003065.3 | 3 | 459 | ||
XM_011534034.1 | XP_011532336.1 | 3 | 190 | |||
GenBank mRNA | AK302541.1 | - | 3 | 234 | ||
BC021862.1 | - | 3 | 396 | |||
BC032564.1 | - | 3 | 443 | |||
BC039843.1 | - | 3 | 363 | |||
BC050564.1 | - | 3 | 388 | 3912713 | ||
BC065253.1 | - | 3 | 428 | |||
BC113465.1 | - | 3 | 407 | 8322517 | ||
BC117213.1 | - | 3 | 407 | 40125764 | ||
BG474088.1 | - | 3 | 460 | |||
U66615.1 | - | 3 | 395 |