Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006946.2 | NP_008877.1 | 29 | 6075 | ||
XM_005274192.4 | XP_005274249.1 | 30 | 6737 | |||
XM_005274193.3 | XP_005274250.1 | 29 | 6511 | |||
XM_006718669.3 | XP_006718732.1 | 28 | 8424 | |||
XM_006718671.3 | XP_006718734.1 | 30 | 6137 | |||
XM_017018174.1 | XP_016873663.1 | 30 | 6829 | |||
XM_017018175.1 | XP_016873664.1 | 30 | 6458 | |||
XM_017018176.1 | XP_016873665.1 | 30 | 6178 | |||
XM_017018177.1 | XP_016873666.1 | 29 | 6367 | |||
XM_017018178.1 | XP_016873667.1 | 29 | 6050 | |||
GenBank mRNA | AB008567.1 | - | 29 | 6075 | ||
AF026487.1 | - | 3 | 308 |