Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SVIL SIRNA Assays ›
Gene Symbol:
SVIL
Gene Name:
supervillin
Gene Aliases:
-
Chromosome Location:
Chr.10: 29457338 - 29737001 on Build GRCh38
UniGene ID:
Hs.499209
Species:
Human
siRNA ID
HSS110394
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SVIL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001323599.1
NP_001310528.1
26
4324
NM_001323600.1
NP_001310529.1
24
3925
NM_003174.3
NP_003165.2
23
3710
NM_021738.2
NP_068506.2
25
5291
XM_005252570.2
XP_005252627.1
29
5314
XM_005252571.3
XP_005252628.1
29
5240
XM_005252573.2
XP_005252630.1
27
5446
XM_011519630.2
XP_011517932.2
26
6081
XM_011519631.2
XP_011517933.2
26
6063
XM_011519632.2
XP_011517934.2
26
5988
XM_011519633.2
XP_011517935.2
24
5937
XM_011519634.1
XP_011517936.1
29
5372
XM_011519635.1
XP_011517937.1
27
5339
XM_011519636.2
XP_011517938.2
26
5229
XM_011519637.2
XP_011517939.2
24
4977
XM_011519638.2
XP_011517940.2
23
4881
XM_017016575.1
XP_016872064.1
27
6159
XM_017016576.1
XP_016872065.1
26
6078
XM_017016577.1
XP_016872066.1
25
6033
XM_017016578.1
XP_016872067.1
25
5133
XM_017016579.1
XP_016872068.1
24
5103
XM_017016580.1
XP_016872069.1
25
5058
XM_017016581.1
XP_016872070.1
22
4851
XM_017016582.1
XP_016872071.1
21
4755
GenBank mRNA
AF051850.1
-
23
3710
AF051851.1
-
19
3360
AF109135.1
-
25
5291
AK302077.1
-
8
996
AK304720.1
-
11
1316
AK304799.1
-
9
1074
AK310272.1
-
8
957
AL117410.1
-
10
1368
JX467682.1
-
20
4442
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
skeletal muscle tissue development
positive regulation of cytokinesis
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0