Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 3 | 228 | ||
NM_003234.3 | NP_003225.2 | 3 | 369 | |||
GenBank mRNA | AB209254.1 | - | 3 | 132 | ||
AK291723.1 | - | 3 | 369 | |||
AK309923.1 | - | 3 | 157 | |||
BC001188.1 | - | 3 | 142 | 3354176 | ||
CF994006.1 | - | 3 | 256 | |||
DA460386.1 | - | 3 | 228 | |||
M11507.1 | - | 3 | 181 | |||
X01060.1 | - | 3 | 349 |