Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323316.1 | NP_001310245.1 | 5, 6 | 770 | ||
NM_001323317.1 | NP_001310246.1 | 3, 4 | 544 | |||
NM_001323318.1 | NP_001310247.1 | 3, 4 | 607 | |||
NM_004613.3 | NP_004604.2 | 4, 5 | 787 | |||
NM_198951.2 | NP_945189.1 | 4, 5 | 787 | |||
XM_011529028.1 | XP_011527330.1 | 5, 6 | 732 | |||
GenBank mRNA | AK058031.1 | - | 4, 5 | 555 | ||
AK291714.1 | - | 4, 5 | 628 | |||
AK295775.1 | - | 3, 4 | 385 | |||
AK300292.1 | - | 3, 4 | 448 | |||
AK314618.1 | - | 4, 5 | 628 | |||
AY675221.1 | - | 3, 4 | 556 | |||
BC003551.1 | - | 4, 5 | 640 | 3544757 | ||
EU794665.1 | - | 4, 5 | 656 | |||
M98478.1 | - | 4, 5 | 688 | |||
S81734.1 | - | 4, 5 | 688 |