Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004615.3 | NP_004606.2 | 2 | 197 | ||
GenBank mRNA | AB062057.1 | - | 2 | 198 | ||
AB102665.1 | - | 2 | 113 | |||
AK293178.1 | - | 3 | 182 | |||
AK293534.1 | - | 4 | 351 | |||
AK293552.1 | - | 3 | 262 | |||
AK312343.1 | - | 2 | 131 | |||
BC018036.1 | - | 2 | 156 | 4801388 | ||
D10653.1 | - | 2 | 137 | |||
D29808.1 | - | 2 | 123 | |||
DB475293.1 | - | 2 | 306 | |||
L10373.1 | - | 2 | 181 |