Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007115.3 | NP_009046.2 | 6 | 785 | ||
GenBank mRNA | AF086484.1 | - | 6 | 637 | ||
AJ419936.1 | - | 6 | 710 | |||
AJ421518.1 | - | 6 | 710 | |||
BC030205.1 | - | 6 | 754 | 5208773 | ||
CA307541.1 | - | 1 | 648 | |||
CA415298.1 | - | 1 | 651 | |||
M31165.1 | - | 6 | 778 |