Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001065.3 | NP_001056.1 | 2, 3 | 492 | ||
GenBank mRNA | A21522.1 | - | 2, 3 | 343 | ||
AK056611.1 | - | 2, 3 | 391 | |||
AK291088.1 | - | 2, 3 | 418 | |||
AK315509.1 | - | 2, 3 | 489 | |||
BC010140.2 | - | 2, 3 | 420 | 4131360 | ||
BF528949.1 | - | 2, 3 | 470 | |||
DA664652.1 | - | 2, 3 | 492 | |||
EU927389.1 | - | 2, 3 | 223 | |||
M33294.1 | - | 2, 3 | 370 | |||
M58286.1 | - | 2, 3 | 375 | |||
M60275.1 | - | 2, 3 | 343 | |||
M63121.1 | - | 2, 3 | 395 |