Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003285.2 | NP_003276.3 | 6 | 1812 | ||
XM_011509949.2 | XP_011508251.1 | 5 | 1421 | |||
XM_017002218.1 | XP_016857707.1 | 7 | 2249 | |||
XM_017002219.1 | XP_016857708.1 | 6 | 2177 | |||
GenBank mRNA | BC129830.1 | - | 4 | 1135 | 40114168 | |
X98085.1 | - | 5 | 1374 | |||
Z67996.1 | - | 4 | 1338 |