Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001068.3 | NP_001059.2 | 28 | 3760 | ||
XM_005265427.3 | XP_005265484.1 | 28 | 4339 | |||
XM_011534057.2 | XP_011532359.1 | 28 | 3749 | |||
GenBank mRNA | AB208879.1 | - | 25 | 3318 | ||
AK296510.1 | - | 29 | 3815 | |||
AK297171.1 | - | 4 | 557 | |||
AK316492.1 | - | 4 | 510 | |||
BC021863.1 | - | 3 | 279 | |||
X53662.1 | - | 5 | 574 | |||
X68060.1 | - | 28 | 3685 | |||
Z15115.1 | - | 14 | 1915 |