Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 10 | 931 | ||
NM_199259.3 | NP_954868.1 | 14 | 1153 | |||
NM_199260.3 | NP_954869.1 | 13 | 1093 | |||
NM_199261.3 | NP_954870.2 | 15 | 1207 | |||
GenBank mRNA | AF007118.1 | - | 15 | 1155 | ||
AF495908.1 | - | 14 | 1101 | |||
AK308254.1 | - | 14 | 1050 | |||
AK314285.1 | - | 15 | 1106 | |||
AL833345.1 | - | 15 | 1114 | |||
AY219886.1 | - | 14 | 1101 | |||
AY219887.1 | - | 13 | 1041 | |||
AY219888.1 | - | 10 | 879 | |||
BC028719.1 | - | 14 | 1136 | 5269598 |