Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142407.2 | NP_001135879.1 | 6 | 551 | ||
NM_001142408.2 | NP_001135880.1 | 7 | 608 | |||
NM_001142417.2 | NP_001135889.1 | 7 | 588 | |||
NM_001142435.2 | NP_001135907.1 | 6 | 571 | |||
NM_001261822.1 | NP_001248751.1 | 6 | 577 | |||
NM_003296.3 | NP_003287.1 | 7 | 614 | |||
XM_005249349.1 | XP_005249406.1 | 7 | 588 | |||
XM_005249350.1 | XP_005249407.1 | 6 | 546 | |||
XM_005249351.3 | XP_005249408.1 | 6 | 566 | |||
XM_005249352.3 | XP_005249409.1 | 6 | 557 | |||
XM_005249353.3 | XP_005249410.1 | 7 | 593 | |||
XM_005249356.1 | XP_005249413.1 | 7 | 591 | |||
XM_011514841.1 | XP_011513143.1 | 7 | 923 | |||
XM_011514842.1 | XP_011513144.1 | 6 | 886 | |||
XM_011514843.2 | XP_011513145.1 | 7 | 593 | |||
XM_011514844.1 | XP_011513146.1 | 7 | 593 | |||
GenBank mRNA | AK223337.1 | - | 7 | 551 | ||
AK292385.1 | - | 7 | 592 | |||
AY289796.1 | - | 7 | 558 | |||
AY292862.1 | - | 6 | 548 | |||
AY292863.1 | - | 6 | 552 | |||
AY773479.1 | - | 7 | 594 | |||
BC022011.2 | - | 7 | 561 | 4826427 | ||
BC107707.1 | - | 6 | 508 | 6618679 | ||
BG718726.1 | - | 6 | 547 | |||
BI562149.1 | - | 6 | 534 | |||
BM552843.1 | - | 6 | 486 | |||
BP369727.1 | - | 1 | 65 | |||
M25532.1 | - | 7 | 537 | |||
X95239.1 | - | 7 | 588 |