Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003355.2 | NP_003346.2 | 5, 6 | 923 | ||
GenBank mRNA | AK025742.1 | - | 5, 6 | 1386 | ||
AK222540.1 | - | 5, 6 | 922 | |||
AK222557.1 | - | 5, 6 | 922 | |||
BC011737.2 | - | 5, 6 | 886 | 4335185 | ||
U76367.1 | - | 3, 4 | 543 | |||
U82819.1 | - | 3, 4 | 631 | |||
U94592.1 | - | 5, 6 | 857 |