Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008389.2 | NP_001008390.1 | 8 | 1819 | ||
NM_001278614.1 | NP_001265543.1 | 9 | 1981 | |||
NM_003361.3 | NP_003352.2 | 8 | 1882 | |||
XM_011545934.1 | XP_011544236.1 | 9 | 1960 | |||
XM_011545935.1 | XP_011544237.1 | 10 | 2019 | |||
XM_011545936.1 | XP_011544238.1 | 10 | 1956 | |||
XM_011545937.1 | XP_011544239.1 | 10 | 1846 | |||
XM_011545938.1 | XP_011544240.1 | 9 | 1878 | |||
XM_011545940.1 | XP_011544242.1 | 9 | 1896 | |||
XM_017023641.1 | XP_016879130.1 | 8 | 1819 | |||
GenBank mRNA | AK055722.1 | - | 9 | 1585 | ||
AK091961.1 | - | 9 | 1813 | |||
AK096043.1 | - | 9 | 1462 | |||
AK127643.1 | - | 9 | 1315 | |||
AK127648.1 | - | 9 | 1578 | |||
BC035975.1 | - | 9 | 1568 | 4556427 | ||
M15881.1 | - | 8 | 1758 | |||
M17778.1 | - | 7 | 1695 |