Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167574.1 | NP_001161046.1 | 4 | 401 | ||
NM_006953.3 | NP_008884.1 | 6 | 764 | |||
GenBank mRNA | AB010116.1 | - | 5 | 641 | ||
AB010637.1 | - | 6 | 737 | |||
AF085808.1 | - | 6 | 769 | |||
BC069544.1 | - | 6 | 769 | 7262218 | ||
BC108900.2 | - | 4 | 397 | 40004236 | ||
BM681088.1 | - | 1 | 296 | |||
CR456608.1 | - | 6 | 739 |