Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003370.3 | NP_003361.1 | 11 | 1362 | ||
XM_005259199.1 | XP_005259256.1 | 11 | 1347 | |||
XM_005259200.1 | XP_005259257.1 | 11 | 1347 | |||
XM_017027200.1 | XP_016882689.1 | 11 | 1344 | |||
GenBank mRNA | AK314812.1 | - | 11 | 1329 | ||
AL556269.3 | - | 9 | 837 | |||
BC026019.1 | - | 11 | 1299 | 3345963 | ||
BC038224.1 | - | 11 | 1280 | 5210128 | ||
KU178381.1 | - | 10 | 1020 | |||
KU178382.1 | - | 11 | 864 | |||
KU178383.1 | - | 11 | 861 | |||
KU178384.1 | - | 10 | 744 | |||
KU178385.1 | - | 8 | 569 | |||
Z46389.1 | - | 11 | 1274 |