Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003381.3 | NP_003372.1 | 5 | 574 | ||
NM_194435.2 | NP_919416.1 | 5 | 571 | |||
XM_005267135.3 | XP_005267192.1 | 5 | 523 | |||
XM_006715562.3 | XP_006715625.1 | 5 | 3477 | |||
GenBank mRNA | AK291959.1 | - | 5 | 523 | ||
AK313950.1 | - | 5 | 574 | |||
BC009794.1 | - | 5 | 576 | 4294373 | ||
M36634.1 | - | 5 | 467 | |||
M54930.1 | - | 3 | 254 |