Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005663.4 | NP_005654.3 | 11 | 1832 | ||
XM_017008589.1 | XP_016864078.1 | 12 | 2653 | |||
GenBank mRNA | AB044549.1 | - | 10 | 1619 | ||
AF101434.1 | - | 11 | 1561 | |||
AF131751.1 | - | 7 | 1082 | |||
AK001304.1 | - | 11 | 1619 | |||
AK094040.1 | - | 10 | 1392 | |||
AK095737.1 | - | 10 | 1875 | |||
AK126056.1 | - | 11 | 2707 | |||
AK222853.1 | - | 11 | 1569 | |||
BC002764.2 | - | 11 | 1572 | 3630594 |