Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167604.1 | NP_001161076.1 | 8, 9 | 860 | ||
NM_001324128.1 | NP_001311057.1 | 6, 7 | 707 | |||
NM_001324131.1 | NP_001311060.1 | 9, 10 | 955 | |||
NM_001324132.1 | NP_001311061.1 | 7, 8 | 771 | |||
NM_001324133.1 | NP_001311062.1 | 8, 9 | 860 | |||
NM_001324134.1 | NP_001311063.1 | 6, 7 | 691 | |||
NM_001324135.1 | NP_001311064.1 | 8, 9 | 860 | |||
NM_001324136.1 | NP_001311065.1 | 8, 9 | 1075 | |||
NM_020383.3 | NP_065116.3 | 8, 9 | 860 | |||
XM_011540134.1 | XP_011538436.1 | 8, 9 | 854 | |||
XM_017016613.1 | XP_016872102.1 | 7, 8 | 710 | |||
XM_017016614.1 | XP_016872103.1 | 9, 10 | 843 | |||
XM_017016615.1 | XP_016872104.1 | 7, 8 | 727 | |||
GenBank mRNA | AF195530.1 | - | 7, 8 | 687 | ||
AF272981.1 | - | 6, 7 | 611 | |||
AK093693.1 | - | 6, 7 | 665 | |||
AK223513.1 | - | 7, 8 | 671 | |||
AK289436.1 | - | 8, 9 | 763 | |||
AK304367.1 | - | 8, 9 | 753 | |||
AL833411.1 | - | 8, 9 | 766 | |||
BC005126.2 | - | 7, 8 | 649 | 3836436 | ||
BC007579.1 | - | 8, 9 | 751 | 3139868 | ||
BC013417.2 | - | 4, 5 | 361 | |||
BX537682.1 | - | 5, 6 | 516 | |||
CR456922.1 | - | 6, 7 | 611 | |||
X95762.1 | - | 6, 7 | 611 |