Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278170.1 | NP_001265099.1 | 4 | 1478 | ||
NM_001278171.1 | NP_001265100.1 | 6 | 1657 | |||
NM_001278173.1 | NP_001265102.1 | 6 | 1613 | |||
NM_001278174.1 | NP_001265103.1 | 6 | 1609 | |||
NM_001278175.1 | NP_001265104.1 | 6 | 1598 | |||
NM_001278176.1 | NP_001265105.1 | 6 | 1614 | |||
NM_001278177.1 | NP_001265106.1 | 5 | 1530 | |||
NM_001278178.1 | NP_001265107.1 | 5 | 1539 | |||
NM_001278179.1 | NP_001265108.1 | 6 | 1654 | |||
NM_001324175.1 | NP_001311104.1 | 6 | 1606 | |||
NM_001324176.1 | NP_001311105.1 | 5 | 1536 | |||
NM_001324177.1 | NP_001311106.1 | 6 | 1614 | |||
NM_001324178.1 | NP_001311107.1 | 4 | 1481 | |||
NM_006954.1 | NP_008885.1 | 5 | 1534 | |||
NM_006974.2 | NP_008905.1 | 5 | 1531 | |||
XM_011519650.2 | XP_011517952.1 | 6 | 1603 | |||
XM_011519651.2 | XP_011517953.1 | 6 | 1600 | |||
XM_011519653.2 | XP_011517955.1 | 4 | 1416 | |||
XM_011519654.2 | XP_011517956.1 | 4 | 1413 | |||
XM_017016617.1 | XP_016872106.1 | 5 | 1497 | |||
GenBank mRNA | AK292844.1 | - | 5 | 1515 | ||
AK303445.1 | - | 4 | 1473 | |||
AY184389.1 | - | 3 | 1353 | |||
BC126119.1 | - | 3 | 1406 | 8991835 | ||
D31763.1 | - | 5 | 1468 |