Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007146.2 | NP_009077.2 | 6 | 1570 | ||
XM_005257643.1 | XP_005257700.1 | 6 | 1567 | |||
XM_005257644.4 | XP_005257701.1 | 7 | 1723 | |||
XM_017025017.1 | XP_016880506.1 | 7 | 1774 | |||
XM_017025018.1 | XP_016880507.1 | 7 | 1626 | |||
GenBank mRNA | AA195154.1 | - | 2 | 427 | ||
BX648862.1 | - | 6 | 863 | |||
D28118.1 | - | 6 | 1453 |