Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYM2 SIRNA Assays ›
Gene Symbol:
ZMYM2
Gene Name:
zinc finger MYM-type containing 2
Gene Aliases:
FIM, MYM, RAMP, SCLL, ZNF198
Chromosome Location:
Chr.13: 19957412 - 20091845 on Build GRCh38
UniGene ID:
Hs.507433
Species:
Human
siRNA ID
HSS111672
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190964.2
NP_001177893.1
11
2054
NM_001190965.2
NP_001177894.1
10
2069
NM_003453.4
NP_003444.1
11
2162
NM_197968.3
NP_932072.1
10
2115
XM_005266520.3
XP_005266577.1
10
2074
XM_011535216.2
XP_011533518.1
11
1744
XM_011535222.1
XP_011533524.1
10
2005
XM_011535223.2
XP_011533525.1
4
4459
XM_011535224.2
XP_011533526.1
4
360
XM_017020728.1
XP_016876217.1
10
2005
XM_017020729.1
XP_016876218.1
10
1980
XM_017020731.1
XP_016876220.1
11
1797
XM_017020732.1
XP_016876221.1
11
1819
XM_017020733.1
XP_016876222.1
12
1846
XM_017020734.1
XP_016876223.1
12
1832
XM_017020735.1
XP_016876224.1
11
1796
XM_017020736.1
XP_016876225.1
11
1968
XM_017020737.1
XP_016876226.1
11
1743
XM_017020738.1
XP_016876227.1
10
2058
XM_017020739.1
XP_016876228.1
11
2085
GenBank mRNA
AF012126.1
-
4
457
AF035374.1
-
8
1360
AF060181.1
-
8
1412
AJ224901.1
-
11
2048
AK226118.1
-
10
2049
AK289741.1
-
10
2040
AK302917.1
-
5
429
BC036372.1
-
10
2115
4823008
BX647944.1
-
10
2069
BX648905.1
-
3
2499
Y13472.1
-
11
2060
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
biological_process
peptidyl-tyrosine phosphorylation
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein tyrosine kinase activity
zinc ion binding
ubiquitin conjugating enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0