Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003694.1 | NP_001003694.1 | 3 | 1229 | ||
NM_001319049.1 | NP_001305978.1 | 3 | 1250 | |||
NM_001319050.1 | NP_001305979.1 | 3 | 1250 | |||
NM_004634.2 | NP_004625.2 | 3 | 1229 | |||
XM_005265449.1 | XP_005265506.1 | 3 | 1250 | |||
XM_005265450.1 | XP_005265507.1 | 3 | 1250 | |||
XM_005265452.1 | XP_005265509.1 | 3 | 1250 | |||
XM_011534101.1 | XP_011532403.1 | 3 | 1250 | |||
XM_011534102.1 | XP_011532404.1 | 3 | 1048 | |||
GenBank mRNA | AK293865.1 | - | 3 | 1250 | ||
AL713696.1 | - | 3 | 1059 | |||
BC053851.1 | - | 3 | 1223 | 6140996 | ||
M91585.1 | - | 3 | 858 |