Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLMAP SIRNA Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.3: 57756221 - 57930013 on Build GRCh38
UniGene ID:
Hs.476432
Species:
Human
siRNA ID
HSS111811
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304420.2
NP_001291349.1
15
1800
NM_001304421.2
NP_001291350.1
13
1686
NM_001304422.2
NP_001291351.1
3
282
NM_001304423.2
NP_001291352.1
3
282
NM_001311178.1
NP_001298107.1
3
282
NM_001311179.1
NP_001298108.1
3
282
NM_007159.4
NP_009090.2
14
1749
XM_005265456.3
XP_005265513.1
17
2687
XM_005265457.3
XP_005265514.1
17
2681
XM_005265458.3
XP_005265515.1
17
2687
XM_005265459.3
XP_005265516.1
16
2636
XM_005265460.3
XP_005265517.1
16
2636
XM_005265461.3
XP_005265518.1
16
2630
XM_005265462.3
XP_005265519.1
16
2624
XM_005265464.3
XP_005265521.1
16
2618
XM_005265465.3
XP_005265522.1
15
2585
XM_005265466.3
XP_005265523.1
15
2579
XM_005265467.3
XP_005265524.1
15
2573
XM_005265468.3
XP_005265525.1
17
2687
XM_005265470.3
XP_005265527.1
14
2522
XM_005265471.3
XP_005265528.1
14
2516
XM_005265472.3
XP_005265529.1
15
2573
XM_006713326.2
XP_006713389.1
15
2579
XM_011534103.1
XP_011532405.1
15
2573
XM_011534104.1
XP_011532406.1
15
2585
XM_011534105.1
XP_011532407.1
14
2522
XM_011534106.1
XP_011532408.1
3
157
XM_017007146.1
XP_016862635.1
17
2681
XM_017007147.1
XP_016862636.1
16
2630
XM_017007148.1
XP_016862637.1
16
2636
XM_017007149.1
XP_016862638.1
16
2630
XM_017007150.1
XP_016862639.1
16
2630
XM_017007151.1
XP_016862640.1
15
2585
XM_017007152.1
XP_016862641.1
15
2579
XM_017007153.1
XP_016862642.1
15
2567
XM_017007154.1
XP_016862643.1
15
2567
XM_017007155.1
XP_016862644.1
17
2681
XM_017007156.1
XP_016862645.1
17
2687
XM_017007157.1
XP_016862646.1
17
2681
XM_017007158.1
XP_016862647.1
14
2522
XM_017007159.1
XP_016862648.1
16
2636
XM_017007160.1
XP_016862649.1
14
2516
XM_017007161.1
XP_016862650.1
16
2630
XM_017007162.1
XP_016862651.1
16
2636
XM_017007163.1
XP_016862652.1
16
2630
XM_017007164.1
XP_016862653.1
15
2579
XM_017007165.1
XP_016862654.1
15
2567
XM_017007166.1
XP_016862655.1
15
2567
XM_017007167.1
XP_016862656.1
14
2522
XM_017007168.1
XP_016862657.1
14
2516
XM_017007169.1
XP_016862658.1
17
2687
XM_017007170.1
XP_016862659.1
3
157
GenBank mRNA
AB046821.1
-
12
990
AF100750.1
-
3
226
AF304450.1
-
3
155
AK124200.1
-
15
1490
AK300106.1
-
3
205
AK302934.1
-
3
157
AK304493.1
-
3
209
AK316436.1
-
3
150
BC114627.1
-
13
1384
40081892
BC115701.1
-
15
1498
40081893
CR627321.1
-
3
139
DB100357.1
-
3
286
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0