Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308217.1 | NP_001295146.1 | 9 | 735 | ||
NM_001308222.1 | NP_001295151.1 | 9 | 1488 | |||
NM_003471.3 | NP_003462.2 | 10 | 942 | |||
NM_172159.3 | NP_751891.1 | 10 | 1625 | |||
NM_172160.2 | NP_751892.1 | 10 | 822 | |||
XM_011513115.2 | XP_011511417.1 | 10 | 535 | |||
XM_011513116.2 | XP_011511418.1 | 8 | 614 | |||
XM_011513117.2 | XP_011511419.1 | 8 | 425 | |||
XM_017007171.1 | XP_016862660.1 | 10 | 612 | |||
XM_017007172.1 | XP_016862661.1 | 10 | 1828 | |||
XM_017007173.1 | XP_016862662.1 | 8 | 529 | |||
GenBank mRNA | AK057059.1 | - | 10 | 1358 | ||
AK127240.1 | - | 10 | 1555 | |||
AK292693.1 | - | 10 | 808 | |||
AK292999.1 | - | 10 | 874 | |||
AK296728.1 | - | 8 | 526 | |||
AK303287.1 | - | 9 | 705 | |||
AY780786.1 | - | 9 | 617 | |||
BC043166.2 | - | 10 | 1575 | 5286833 | ||
L39833.1 | - | 10 | 813 | |||
L47665.1 | - | 10 | 811 | |||
U16953.1 | - | 10 | 792 | |||
U33428.1 | - | 10 | 731 |