Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306074.1 | NP_001293003.1 | 4 | 898 | ||
NM_001306075.1 | NP_001293004.1 | 4 | 886 | |||
NM_006527.3 | NP_006518.1 | 5 | 1003 | |||
GenBank mRNA | AK091735.1 | - | 4 | 463 | ||
AK093314.1 | - | 5 | 476 | |||
AK315635.1 | - | 5 | 574 | |||
BC014908.1 | - | 5 | 572 | 3908584 | ||
BC015703.1 | - | 5 | 572 | 3907984 | ||
BT007433.1 | - | 5 | 450 | |||
DB031118.1 | - | 5 | 476 | |||
U75679.1 | - | 5 | 565 | |||
Z71188.1 | - | 5 | 554 |