Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018412.3 | NP_060882.2 | 7 | 996 | ||
NM_021908.2 | NP_068708.1 | 8 | 1065 | |||
GenBank mRNA | AF234882.1 | - | 7 | 995 | ||
AF234883.1 | - | 8 | 1064 | |||
AF415174.1 | - | 5 | 423 | |||
AJ277291.1 | - | 6 | 631 | |||
AK093212.1 | - | 8 | 1245 | |||
AK292372.1 | - | 7 | 822 | |||
AK297510.1 | - | 8 | 942 | |||
AK297861.1 | - | 8 | 773 | |||
AK298540.1 | - | 7 | 818 | |||
AY009152.1 | - | 8 | 1056 | |||
AY009153.1 | - | 7 | 987 | |||
BC030954.1 | - | 8 | 801 | |||
BC075855.1 | - | 8 | 1065 | 5562032 | ||
CR936656.1 | - | 2 | 206 | |||
KJ534947.1 | - | 8 | 851 |