Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006767.3 | NP_006758.2 | 15, 16 | 1899 | ||
GenBank mRNA | AK296347.1 | - | 14, 15 | 1780 | ||
AK313502.1 | - | 10, 11 | 1301 | |||
BC026214.1 | - | 15, 16 | 1801 | 4399716 | ||
CT841521.1 | - | 15, 16 | 1815 | |||
D38496.1 | - | 15, 16 | 1794 |