Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142391.2 | NP_001135863.1 | 4 | 1278 | ||
NM_001142392.2 | NP_001135864.1 | 3 | 1365 | |||
NM_019848.4 | NP_062822.1 | 2 | 1600 | |||
XM_005277913.4 | XP_005277970.1 | 2 | 2022 | |||
XM_006724847.3 | XP_006724910.1 | 2 | 1543 | |||
XM_006724848.3 | XP_006724911.1 | 3 | 1456 | |||
XM_011531200.2 | XP_011529502.2 | 2 | 1575 | |||
XM_011531201.2 | XP_011529503.1 | 2 | 1279 | |||
GenBank mRNA | BC004966.1 | - | 4 | 1202 | 3503242 | |
BM981524.1 | - | 1 | 576 |