Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013836.1 | NP_001013858.1 | 13 | 1591 | ||
NM_001013837.1 | NP_001013859.1 | 13 | 1588 | |||
NM_001304523.1 | NP_001291452.1 | 12 | 1412 | |||
NM_001304524.1 | NP_001291453.1 | 11 | 1250 | |||
NM_003550.2 | NP_003541.2 | 13 | 1628 | |||
XM_005249877.1 | XP_005249934.1 | 11 | 1307 | |||
XM_011515567.1 | XP_011513869.1 | 11 | 1335 | |||
XM_011515568.2 | XP_011513870.1 | 11 | 1335 | |||
XM_011515569.2 | XP_011513871.2 | 2 | 362 | |||
XM_011515571.1 | XP_011513873.1 | 4 | 601 | |||
XM_017012690.1 | XP_016868179.1 | 13 | 1806 | |||
XM_017012691.1 | XP_016868180.1 | 9 | 1206 | |||
GenBank mRNA | AF083811.1 | - | 13 | 1574 | ||
AF123318.1 | - | 11 | 1325 | |||
AK090959.1 | - | 11 | 1247 | |||
AK310481.1 | - | 11 | 1216 | |||
AK310831.1 | - | 13 | 1588 | |||
AK314227.1 | - | 12 | 1409 | |||
BC009964.2 | - | 12 | 1538 | 4299982 | ||
U33822.1 | - | 12 | 1501 |