Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004426.1 | NP_001004426.1 | 2, 3 | 337 | ||
NM_001199562.1 | NP_001186491.1 | 2, 3 | 292 | |||
NM_003560.2 | NP_003551.2 | 2, 3 | 337 | |||
XM_005261764.2 | XP_005261821.1 | 2, 3 | 312 | |||
XM_005261765.1 | XP_005261822.1 | 2, 3 | 309 | |||
XM_005261766.1 | XP_005261823.1 | 2, 3 | 404 | |||
XM_006724332.3 | XP_006724395.1 | 2, 3 | 313 | |||
XM_011530422.1 | XP_011528724.1 | 2, 3 | 293 | |||
XM_011530426.2 | XP_011528728.1 | 2, 3 | 312 | |||
XM_017028981.1 | XP_016884470.1 | 2, 3 | 404 | |||
XM_017028982.1 | XP_016884471.1 | 2, 3 | 309 | |||
XM_017028985.1 | XP_016884474.1 | 2, 3 | 268 | |||
XM_017028986.1 | XP_016884475.1 | 2, 3 | 312 | |||
XM_017028987.1 | XP_016884476.1 | 2, 3 | 312 | |||
XM_017028988.1 | XP_016884477.1 | 2, 3 | 312 | |||
GenBank mRNA | AF064594.1 | - | 2, 3 | 337 | ||
AF102988.1 | - | 1, 2 | 201 | |||
AF102989.1 | - | 1, 2 | 201 | |||
AK096521.1 | - | 2, 3 | 287 | |||
AK290825.1 | - | 2, 3 | 294 | |||
AK291212.1 | - | 2, 3 | 323 | |||
AK300459.1 | - | 2, 3 | 293 | |||
AL080187.1 | - | 2, 3 | 337 | |||
BC036742.1 | - | 2, 3 | 331 | 5166749 | ||
BC051904.1 | - | 2, 3 | 273 | 5175768 | ||
CR456543.1 | - | 2, 3 | 265 |