Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170750.1 | NP_001164221.1 | 3 | 452 | ||
NM_001170752.1 | NP_001164223.1 | 4 | 512 | |||
NM_006307.4 | NP_006298.1 | 4 | 512 | |||
XM_017029893.1 | XP_016885382.1 | 4 | 451 | |||
GenBank mRNA | AK125542.1 | - | 3 | 433 | ||
AK289430.1 | - | 4 | 459 | |||
AK293127.1 | - | 4 | 489 | |||
BC020684.1 | - | 4 | 467 | 4731946 | ||
DA422854.1 | - | 3 | 425 | |||
DC388035.1 | - | 4 | 512 | |||
EU668359.1 | - | 4 | 418 | |||
U61374.1 | - | 4 | 418 | |||
U78093.1 | - | 4 | 419 |