Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032296.3 | NP_001027467.2 | 8 | 1377 | ||
NM_001286649.1 | NP_001273578.1 | 7 | 1320 | |||
NM_003576.4 | NP_003567.2 | 8 | 1321 | |||
XM_005254079.4 | XP_005254136.1 | 8 | 1198 | |||
XM_017020794.1 | XP_016876283.1 | 8 | 1494 | |||
XM_017020795.1 | XP_016876284.1 | 8 | 1156 | |||
GenBank mRNA | AF024636.1 | - | 8 | 1090 | ||
AF083420.1 | - | 8 | 1193 | |||
AF339826.1 | - | 7 | 798 | |||
AK299140.1 | - | 7 | 1037 | |||
BC035578.1 | - | 8 | 1325 | 5539375 | ||
BC065378.1 | - | 8 | 1321 | 6598075 | ||
BT019619.1 | - | 8 | 1012 | |||
CR542227.1 | - | 8 | 1012 | |||
EU668350.1 | - | 8 | 1368 |